Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module

Catalog No. 13645594 Shop All Thermo Scientific Products
Change view
Click to view available options
pH Range:
-2.000 to 20.000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
13-645-594 -2.000 to 20.000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-645-594 Supplier Thermo Scientific™ Supplier No. VSTARISE
  • $2,440.00
    $1,975.65 / Each of 1

    Save $464.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Add pH, ion concentration, mV, ORP and temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module

Add pH, ion concentration, mV, ORP and temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Versa Star pH/ISE module offers up to six point pH and ISE calibrations and advanced ISE features including incremental techniques (single and double known addition and subtraction, analate addition and subtraction), low range stability and electrode ID with calibration verification.

Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter's four channels at any time.

Orion Versa Star pH/ISE Measurement Module:

  • Measure pH, ion concentration, mV, relative mV or ORP with temperature
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Perform up to a six point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Incremental techniques include single known addition, double known addition, single known subtraction, double known subtraction, analate addition and analate subtraction
  • Advanced ISE features include non-linear selectable auto-blank, low range stability and electrode ID with calibration verification
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH or ion concentration reading
  • Module offers a 3-year replacement warranty

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy (pH) ±0.002 pH
Accuracy (ISE) ±0.2 mV or ±0.05 of reading, whichever is greater
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Advanced ISE Functions Timed endpoint, linear point to point, non-linear selectable auto-blank, low concentration range stability, incremental techniques including single known addition and subtraction, double known addition and subtraction, single analyte addition and subtraction
Accuracy (Temperature) ±0.1°C
Alarm Output Set point alarm, high/low limit alarm, calibration due alarm
Calibration 1 point (mV mode)
Calibration Points (ISE) 1 to 6
Calibration Points (pH) 1 to 6
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Description Orion VERSA STAR pH/mV/ISE/Temperature Module
Type pH/ISE Measurement Module
Dimensions (L x W x H) 12 x 3 x 9.5 cm (4.7 x 1.2 x 3.8 in.)
Inputs BNC (pH or ISE), 8-pin Mini-DIN (temperature), reference pin-tip
Includes VERSA STAR pH/mV/ISE/Temperature Module
Isopotential Point 7.000pH (adjustable), 1.000 ISE (adjustable)
IP Rating IP-54
Measurement Modes pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
Methods 10 per channel with password protection
Model Orion Versa Star pH/ISE/mV/RmV/ORP/Temperature Measurement Module
pH Range -2.000 to 20.000
Probe Type pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe (purchase separately)
Range (ISE) 0 to 19999 in ppm, M, mg/L, %, ppb or unitless
Range (mV) ±2000.0mV
Range (ORP) ±2000.0mV
Relative Humidity Range 5 to 85 %, non-condensing
Resolution (ISE) Up to 4 significant digits
Resolution (mV) 0.1mV
Resolution (pH) 0.1, 0.01, 0.001
Resolution (Temperature) 0.1°C
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Operating 41°F to 113°F
Temperature (Metric) Range -5°C to 105deg;C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature (Metric) Operating 5°C to 45°C
Temperature Selection Manual or automatic with ATC temperature probe
Weight (English) 0.5 lb.
Warranty 3 year module warranty
Weight (Metric) 0.2 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.