Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ Versa Star pH / LogR Measurement Module

Catalog No. 13645595 Shop All Thermo Scientific Products
Change view
Click to view available options
pH Range:
-2.000 to 20.000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
13-645-595 -2.000 to 20.000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-645-595 Supplier Thermo Scientific™ Supplier No. VSTARLR
  • $1,941.00
    $1,493.65 / Each of 1

    Save $447.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Add pH, mV, ORP and LogR temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / LogR Measurement Module

Add pH, mV, ORP and LogR temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / LogR Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Orion™ Versa Star pH/LogR module offers LogR temperature technology that uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location – ideal for small sample volumes and samples that are difficult to clean.

Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter’s four channels at any time.

Includes Orion Versa Star pH/LogR Measurement Module:

  • pH, mV, relative mV or ORP with temperature using LogR technology, ATC probe or manual input
  • LogR technology uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location – ideal for small sample volumes and samples that are difficult to clean
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
  • Module offers a 3-year replacement warranty

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy (pH) ±0.002 pH
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (Temperature) ±0.1°C
Alarm Output Set point alarm, high/low limit alarm, calibration due alarm
Calibration pH with calibration editing option, relative mV (RmV), ORP, LogR temperature and temperature
Calibration Points 1 to 6
Description Orion Versa Star pH/LogR measurement module
Type pH/LogR Measurement Module
Dimensions (L x W x H) 12 x 6 x 9.5 cm (4.7 x 2.4 x 3.8 in.)
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Includes Versa Star pH/LogR measurement module for use with Versa Star Pro meters to measure pH, mV, relative mV, ORP and temperature using LogR technology, ATC probe or manual entry
Isopotential Point 7.000pH (adjustable)
IP Rating IP-54
Measurement Modes pH with LogR temperature, mV, relative mV (RmV) or ORP with temperature
Methods 10 per channel with password protection
Model Orion Versa Star pH/LogR/mV/RmV/ORP/Temperature Measurement Module
pH Range -2.000 to 20.000
Probe Type pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (purchase separately)
Range (mV) ±2000.0 mV
Range (ORP) ±2000.0mV
Relative Humidity Range 5 to 85%, non-condensing
Resolution (mV) 0.1mV
Resolution (pH) 0.1, 0.01, 0.001
Resolution (Temperature) 0.1°F/°C
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Range 22.0°F to 221.0°F
Temperature (English) Operating 41°F to 113°F
Temperature (Metric) Range -5°C to 105deg;C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature (Metric) Operating 5°C to 45°C
Temperature Selection Manual or automatic with ATC temperature probe, automatic with pH electrode using LogR mode
Weight (English) 0.5 lb.
Warranty 3 year module warranty
Weight (Metric) 0.2 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.