Learn More
Medchemexpress LLC Orismilast | 1353546-86-7 | 99.9% | 510.29 | C19H15Cl2F2NO7S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11796010MG
Encompass_Preferred
Orismilast is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor used for research into inflammatory diseases. It demonstrates potent inhibition of PDE4B and PDE4D subtypes and is provided as a solid research reagent for preclinical in vitro and in vivo studies. Not for human use.
  • Selective inhibition of PDE4B and PDE4D, targeting inflammatory pathways.
  • Orally active small molecule suitable for preclinical pharmacology studies.
  • High chemical purity (≈99.9%) and defined molecular weight (510.29 g/mol).
  • Molecular formula C19H15Cl2F2NO7S; suitable for screening and mechanistic studies.
  • Supplied as a solid research reagent; follow safety and handling guidelines.
Catalog No. 50-002-27000