Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orismilast | 1353546-86-7 | 99.9% | 510.29 g/mol | C19H15Cl2F2NO7S | 100 MG
SDP

Catalog No. 5000227697
Encompass_Preferred

Orismilast (LEO-32731) is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor used in research on inflammatory diseases. It exhibits potent inhibition of PDE4B and PDE4D splice variants and is supplied as a high-purity research compound for in vitro and preclinical studies.

  • Selective PDE4 inhibitor with activity against PDE4B and PDE4D.
  • Purity 99.9%.
  • Molecular weight 510.29 g/mol; formula C19H15Cl2F2NO7S.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Powder storage: -20°C for long-term stability; aliquot to avoid freeze-thaw.
  • Intended for in vitro and preclinical research use only.

Catalog No. 50-002-27697 Supplier Medchemexpress LLC Supplier No. HY117960100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orismilast (LEO-32731) is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor used in research on inflammatory diseases. It exhibits potent inhibition of PDE4B and PDE4D splice variants and is supplied as a high-purity research compound for in vitro and preclinical studies.

  • Selective PDE4 inhibitor with activity against PDE4B and PDE4D.
  • Purity 99.9%.
  • Molecular weight 510.29 g/mol; formula C19H15Cl2F2NO7S.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Powder storage: -20°C for long-term stability; aliquot to avoid freeze-thaw.
  • Intended for in vitro and preclinical research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.