Learn More
Medchemexpress LLC Orismilast | 1353546-86-7 | 99.9% | 510.29 g·mol⁻¹ | C19H15Cl2F2NO7S | 5 MG
SDP
Catalog No. 5000226762
Encompass_Preferred
Orismilast is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor used in preclinical research on inflammatory diseases. It demonstrates potent activity against PDE4B and PDE4D subtype splice variants and is supplied as a high-purity research compound.
  • Selective phosphodiesterase 4 (PDE4) inhibitor.
  • Orally active compound suitable for inflammatory disease research.
  • Potent inhibition of PDE4B and PDE4D subtype splice variants.
  • High purity solid supplied in a 5 mg package.
  • Molecular weight 510.29 g·mol⁻¹; formula C19H15Cl2F2NO7S.
Catalog No. 50-002-26762 Supplier Medchemexpress LLC Supplier No. HY1179605MG
Orismilast is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor used in preclinical research on inflammatory diseases. It demonstrates potent activity against PDE4B and PDE4D subtype splice variants and is supplied as a high-purity research compound.
  • Selective phosphodiesterase 4 (PDE4) inhibitor.
  • Orally active compound suitable for inflammatory disease research.
  • Potent inhibition of PDE4B and PDE4D subtype splice variants.
  • High purity solid supplied in a 5 mg package.
  • Molecular weight 510.29 g·mol⁻¹; formula C19H15Cl2F2NO7S.