Learn More
Medchemexpress LLC Orismilast (LEO-32731) | 1353546-86-7 | 99.9% | 510.29 | C19H15Cl2F2NO7S | 50 MG
SDP
Catalog No. 5000226349
Encompass_Preferred
Orismilast is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor provided as a solid research reagent for in vitro and in vivo studies of inflammatory and dermatological disease pathways. It is supplied with high purity and characterized physical data for laboratory use only.
  • Selective PDE4 inhibitor for inflammatory research.
  • High purity (99.88%) solid suitable for analytical and biological assays.
  • Molecular weight 510.29 and formula C19H15Cl2F2NO7S for precise dosing and calculations.
  • Available in small-scale packages including 50 mg for preclinical studies.
  • Stable when stored frozen; follow manufacturer storage recommendations.
Catalog No. 50-002-26349 Supplier Medchemexpress LLC Supplier No. HY11796050MG
Orismilast is an orally active, selective phosphodiesterase 4 (PDE4) inhibitor provided as a solid research reagent for in vitro and in vivo studies of inflammatory and dermatological disease pathways. It is supplied with high purity and characterized physical data for laboratory use only.
  • Selective PDE4 inhibitor for inflammatory research.
  • High purity (99.88%) solid suitable for analytical and biological assays.
  • Molecular weight 510.29 and formula C19H15Cl2F2NO7S for precise dosing and calculations.
  • Available in small-scale packages including 50 mg for preclinical studies.
  • Stable when stored frozen; follow manufacturer storage recommendations.