Learn More
TARGETMOL CHEMICALS INC Oritavancin diphosphate 10MG
SDP
Catalog No. 502877519
Encompass_Preferred
Also available in 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Oritavancin diphosphate (LY333328 diphosphate) is a novel semisynthetic glycopeptide antibiotic developed for treating serious Gram-positive bacterial infections. Purity 100%
Catalog No. 50-287-7519 Supplier TARGETMOL CHEMICALS INC Supplier No. T269110MG
Also available in 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Oritavancin diphosphate (LY333328 diphosphate) is a novel semisynthetic glycopeptide antibiotic developed for treating serious Gram-positive bacterial infections. Purity 100%