Learn More
Medchemexpress LLC Oritinib | 2035089-28-0 | 99.6% | 539.67 g/mol | C31H37N7O2 | 5 MG
SDP
Catalog No. 5000221412
Encompass_Preferred
Oritinib is an irreversible, third-generation EGFR tyrosine kinase inhibitor developed to overcome T790M-mediated resistance. Supplied as a high-purity solid research compound, it is used in preclinical biochemical and cellular studies to interrogate EGFR signaling and resistance mechanisms.
  • Irreversible third-generation EGFR inhibitor.
  • Active against T790M resistance mutations.
  • High chemical purity (99.6%).
  • Provided as a solid suitable for dosing and formulation.
  • Applicable to biochemical and cellular assays.
Catalog No. 50-002-21412 Supplier Medchemexpress LLC Supplier No. HY1399205MG
Oritinib is an irreversible, third-generation EGFR tyrosine kinase inhibitor developed to overcome T790M-mediated resistance. Supplied as a high-purity solid research compound, it is used in preclinical biochemical and cellular studies to interrogate EGFR signaling and resistance mechanisms.
  • Irreversible third-generation EGFR inhibitor.
  • Active against T790M resistance mutations.
  • High chemical purity (99.6%).
  • Provided as a solid suitable for dosing and formulation.
  • Applicable to biochemical and cellular assays.