Learn More
Selleck Chemical LLC Orlistat 100mg 96829-58-2 Ro 18-0647, Tetrahydrolipstatin
SDP
Supplier:  Selleck Chemical LLC S1629100MG
Encompass_Preferred
Orlistat (Ro 18-0647, Tetrahydrolipstatin) is a general lipase inhibitor with IC50 of 122 ng/ml for PL from human duodenal juice. Orlistat treatment reduces proliferation, induces apoptosis and arrests cell cycle. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-580-9