Learn More
Cayman Chemical Orlistat-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1356930-46-5, 1 mg
SDP
Catalog No. 502947441
Encompass_Preferred
An internal standard for the quantification of orlistat by GC- or LC-MS
Catalog No. 50-294-7441 Supplier Cayman Chemical Supplier No. 343611 MG
An internal standard for the quantification of orlistat by GC- or LC-MS