Learn More
TARGETMOL CHEMICALS INC ORM-10103 5MG
SDP
Catalog No. 502577427
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ORM-10103 is a novel potent and selective inhibitor of the Na+/Ca2+ exchanger (NCX).ORM-10103 inhibited NCX currents with estimated IC50 values of 55 and 67 nM at -80 and 20 mV respectively and reduced early and delayed postdepolarization in canine hearts. purity: 98%
Catalog No. 50-257-7427 Supplier TARGETMOL CHEMICALS INC Supplier No. T123215MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ORM-10103 is a novel potent and selective inhibitor of the Na+/Ca2+ exchanger (NCX).ORM-10103 inhibited NCX currents with estimated IC50 values of 55 and 67 nM at -80 and 20 mV respectively and reduced early and delayed postdepolarization in canine hearts. purity: 98%