Learn More
Medchemexpress LLC Orm-10962 | 763926-98-3 | 99.0% | 459.54 g/mol | C27H29N3O4 | 10MG
SDP
Catalog No. 5000237498
Encompass_Preferred
ORM-10962 is a potent, highly selective inhibitor of the sodium-calcium exchanger (NCX) used as a research tool in cellular electrophysiology and cardiovascular studies. It exhibits low-nanomolar activity against both reverse and forward modes of NCX and has a defined chemical composition (C27H29N3O4, MW 459.54 g/mol).
  • Inhibits Na+/Ca2+ exchanger reverse mode (IC50 67 nM) and forward mode (IC50 55 nM).
  • Low-nanomolar potency suitable for electrophysiology assays.
  • Defined chemical formula C27H29N3O4 and molecular weight 459.54 g/mol.
  • High purity (99.0%) as reported by third-party suppliers.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
Catalog No. 50-002-37498 Supplier Medchemexpress LLC Supplier No. HY12378510MG
ORM-10962 is a potent, highly selective inhibitor of the sodium-calcium exchanger (NCX) used as a research tool in cellular electrophysiology and cardiovascular studies. It exhibits low-nanomolar activity against both reverse and forward modes of NCX and has a defined chemical composition (C27H29N3O4, MW 459.54 g/mol).
  • Inhibits Na+/Ca2+ exchanger reverse mode (IC50 67 nM) and forward mode (IC50 55 nM).
  • Low-nanomolar potency suitable for electrophysiology assays.
  • Defined chemical formula C27H29N3O4 and molecular weight 459.54 g/mol.
  • High purity (99.0%) as reported by third-party suppliers.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.