Learn More
Medchemexpress LLC Orm-10962 | 763926-98-3 | MFCD32067938 | 99.5% | 459.54 g/mol | C27H29N3O4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12378550MG
Encompass_Preferred
ORM-10962 is a potent, highly selective inhibitor of the sodium-calcium exchanger (NCX) used for research into cardiac electrophysiology and cellular calcium handling. It inhibits NCX in both reverse and forward modes with low-nanomolar potency and is supplied as a light yellow solid with high reported purity.
  • Potent NCX inhibition (IC50 ~55-67 nM).
  • High reported purity (>99%).
  • Good solubility in DMSO (≈250 mg/mL).
  • Supplied as a light yellow solid for solid-sample handling.
  • Storage-stable when kept frozen as recommended.
  • Suitable for electrophysiology and cellular assays investigating NCX function.
Catalog No. 50-002-37533