Learn More
Apexbio Technology LLC Ornidazole 16773-42-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063764
Encompass_Preferred
Ornidazole a 5-nitroimidazole derivative exhibits antibacterial and antiprotozoal activities by interfering with DNA synthesis in anaerobic microorganisms through the reduction of its nitro group forming cytotoxic intermediates In experimental assays Ornidazole demonstrates inhibitory activity against diverse anaerobic bacterial strains including Bacteroides and Clostridium species and protozoan pathogens such as Giardia lamblia and Trichomonas vaginalis typically showing IC50 values ranging from micromolar to low millimolar concentrations This compound is widely utilized in microbiology and infectious disease research to elucidate anaerobic microbial drug sensitivity pathogen resistance mechanisms and protozoan infection biology
Catalog No. 50-000-63764 Supplier Apexbio Technology LLC Supplier No. B19965.1
Ornidazole a 5-nitroimidazole derivative exhibits antibacterial and antiprotozoal activities by interfering with DNA synthesis in anaerobic microorganisms through the reduction of its nitro group forming cytotoxic intermediates In experimental assays Ornidazole demonstrates inhibitory activity against diverse anaerobic bacterial strains including Bacteroides and Clostridium species and protozoan pathogens such as Giardia lamblia and Trichomonas vaginalis typically showing IC50 values ranging from micromolar to low millimolar concentrations This compound is widely utilized in microbiology and infectious disease research to elucidate anaerobic microbial drug sensitivity pathogen resistance mechanisms and protozoan infection biology