Learn More
Cayman Chemical OrnIpresInacetate 10mg
SDP
Catalog No. 502909692
Encompass_Preferred
A vasopressin V1A receptor agonist; selectively induces reporter gene expression in HEK293 cells expressing vasopressin V1A and V2 receptors in reporter assays (EC50s = 0.69 and 0.45 nM, respectively, for the human receptors) over cells expressing vasopressin V1B receptors; selective for vasopressin V1A and V2 receptors over the vasopressin V1B receptor and oxytocin receptor (EC50s = 7.5 and 71 nM, respectively); increases arterial blood pressure in rats at 0.1 nmol/kg
Catalog No. 50-290-9692 Supplier Cayman Chemical Supplier No. 3671610 MG
A vasopressin V1A receptor agonist; selectively induces reporter gene expression in HEK293 cells expressing vasopressin V1A and V2 receptors in reporter assays (EC50s = 0.69 and 0.45 nM, respectively, for the human receptors) over cells expressing vasopressin V1B receptors; selective for vasopressin V1A and V2 receptors over the vasopressin V1B receptor and oxytocin receptor (EC50s = 7.5 and 71 nM, respectively); increases arterial blood pressure in rats at 0.1 nmol/kg