Learn More
Medchemexpress LLC Ornipressin acetate | 914453-98-8 | 99.6% | 1042.19 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0083A10MG
Encompass_Preferred
Ornipressin acetate (POR-8) is a potent vasoconstrictor and hemostatic agent that acts as a vasopressin agonist specific for the V1 receptor. It can be utilized as a local vasoconstrictor, can reverse hypotension associated with combined general/epidural anesthesia, and possesses antidiuretic activity. Ornipressin acetate also demonstrates the ability to decrease renal vascular resistance and increase renal blood flow in renal failure models.
  • Potent vasoconstrictor
  • Hemostatic and renal agent
  • Vasopressin V1 receptor agonist
  • Antidiuretic activity
  • Decreases renal vascular resistance
  • Increases renal blood flow in renal failure models
Catalog No. 50-004-26326