Learn More
Medchemexpress LLC Ornithine carbamoyltransferase/OTC antibody (YA1230) | 100 UL
SDP
Catalog No. 5000440943
Encompass_Preferred
Ornithine Carbamoyltransferase/OTC Antibody (YA1230) is a Rabbit-derived, non-conjugated IgG monoclonal antibody developed by MedChemExpress. It targets Ornithine Carbamoyltransferase/OTC, an enzyme crucial for the urea cycle. This antibody is suitable for immunohistochemistry-paraffin (IHC-P) applications and is reactive with human samples. It has an observed and predicted band size of 36 kDa.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Targets Ornithine Carbamoyltransferase/OTC
  • Reactive with human samples
  • Suitable for IHC-P application
  • Observed and predicted band size of 36 kDa
  • Stored at -20°C for 1 year
  • Supplied in PBS, pH 7.4
Catalog No. 50-004-40943 Supplier Medchemexpress LLC Supplier No. HYP81485100UL
Ornithine Carbamoyltransferase/OTC Antibody (YA1230) is a Rabbit-derived, non-conjugated IgG monoclonal antibody developed by MedChemExpress. It targets Ornithine Carbamoyltransferase/OTC, an enzyme crucial for the urea cycle. This antibody is suitable for immunohistochemistry-paraffin (IHC-P) applications and is reactive with human samples. It has an observed and predicted band size of 36 kDa.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Targets Ornithine Carbamoyltransferase/OTC
  • Reactive with human samples
  • Suitable for IHC-P application
  • Observed and predicted band size of 36 kDa
  • Stored at -20°C for 1 year
  • Supplied in PBS, pH 7.4