Learn More
Medchemexpress LLC Ornithine Carbamoyltransferase/OTC Antibody (YA1230)(PBS only) | 10 UL
SDP
Catalog No. 5000441203
Encompass_Preferred
This Rabbit-derived IgG monoclonal antibody, targeting Ornithine Carbamoyltransferase/OTC, is designed for research applications. The target protein, a mitochondrial matrix enzyme, plays a crucial role in the urea cycle, detoxifying ammonia into urea. Dysfunction of this enzyme leads to ornithine transcarbamylase deficiency and hyperammonemia.
  • Derived from rabbit host
  • Reacts with human
  • Non-conjugated
  • Predicted and observed molecular weight: 36 kDa
  • Affinity purified
  • Supplied in PBS, pH 7.4
  • Recommended for IHC-P applications at 1:100-1:200 dilution
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
Catalog No. 50-004-41203 Supplier Medchemexpress LLC Supplier No. HYP8148510UL
This Rabbit-derived IgG monoclonal antibody, targeting Ornithine Carbamoyltransferase/OTC, is designed for research applications. The target protein, a mitochondrial matrix enzyme, plays a crucial role in the urea cycle, detoxifying ammonia into urea. Dysfunction of this enzyme leads to ornithine transcarbamylase deficiency and hyperammonemia.
  • Derived from rabbit host
  • Reacts with human
  • Non-conjugated
  • Predicted and observed molecular weight: 36 kDa
  • Affinity purified
  • Supplied in PBS, pH 7.4
  • Recommended for IHC-P applications at 1:100-1:200 dilution
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles