Learn More
Stoelting™ Orofacial Pain Assessment Device
SDP
Catalog No. 10000377 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
OPAD Additional cage
OPAD system
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-377 OPAD Additional cage
10-000-376 OPAD system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-377 Supplier Stoelting™ Supplier No. 67501
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Orofacial Pain Assessment Device provides an automated measurement of hot or cold or mechanical orofacial pain assessment in the trigeminal nerve in rats and mice. Using an operant behavior conflict paradigm, rodents voluntarily decide between receiving a reward or avoiding an aversive stimulus.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat
Model OPAD Additional cage