Learn More
Apexbio Technology LLC Orotic acid 65-86-1 500mg
SDP
Catalog No. 502240559
Encompass_Preferred
Orotic acid (CAS 65-86-1) is a naturally occurring pyrimidine carboxylic acid that functions as a metabolic intermediate in the de novo biosynthesis pathway of pyrimidine nucleotides It participates in the transformation of dihydroorotate to uridine monophosphate (UMP) a precursor for RNA and DNA synthesis Orotic acid is commonly utilized in research to investigate disorders of pyrimidine metabolism such as orotic aciduria and urea cycle deficiencies Its accumulation serves as a diagnostic marker in metabolic studies and disease models providing insights into nucleotide biosynthesis and related metabolic pathologies
This item is not returnable. View return policy
Catalog No. 50-224-0559 Supplier Apexbio Technology LLC Supplier No. B1147500
Orotic acid (CAS 65-86-1) is a naturally occurring pyrimidine carboxylic acid that functions as a metabolic intermediate in the de novo biosynthesis pathway of pyrimidine nucleotides It participates in the transformation of dihydroorotate to uridine monophosphate (UMP) a precursor for RNA and DNA synthesis Orotic acid is commonly utilized in research to investigate disorders of pyrimidine metabolism such as orotic aciduria and urea cycle deficiencies Its accumulation serves as a diagnostic marker in metabolic studies and disease models providing insights into nucleotide biosynthesis and related metabolic pathologies