Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orotic acid hydrate | 50887-69-9 | MFCD00149398 | 99.3% | 174.11 g/mol | C5H4N2O4·H2O | 25 G
SDP

Catalog No. 5000303058
Encompass_Preferred

Orotic acid hydrate is an endogenous metabolite supplied as a research-grade reagent for biochemical and metabolic studies. It is provided as a characterized solid with supporting analytical documentation (COA, LC-MS, SDS) for use as a reference standard and in pathway investigations.

  • High purity: 99.32%.
  • Molecular weight: 174.11 g/mol.
  • Molecular formula: C5H4N2O4·H2O.
  • Documented analytical data: COA and LC-MS available.
  • Soluble in DMSO; solution formats available for convenience.

Catalog No. 50-003-03058 Supplier Medchemexpress LLC Supplier No. 25GHYW016812
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Orotic acid hydrate is an endogenous metabolite supplied as a research-grade reagent for biochemical and metabolic studies. It is provided as a characterized solid with supporting analytical documentation (COA, LC-MS, SDS) for use as a reference standard and in pathway investigations.

  • High purity: 99.32%.
  • Molecular weight: 174.11 g/mol.
  • Molecular formula: C5H4N2O4·H2O.
  • Documented analytical data: COA and LC-MS available.
  • Soluble in DMSO; solution formats available for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.