Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orotic acid hydrate, 98%

Catalog No. AAA1859418
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1859418 50 g
AAA1859430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1859418 Supplier Thermo Scientific Chemicals Supplier No. A1859418
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 50887-69-9 | C5H4N2O4 | 156.10 g/mol

Orotic acid hydrate is used in cosmetic and food industry. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orotic acid hydrate is used in cosmetic and food industry. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in water (1.7 mg/ml), and ammonium hydroxide. Insoluble in chloroform.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50887-69-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00006027
InChI Key PXQPEWDEAKTCGB-UHFFFAOYSA-N
Synonym orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
PubChem CID 16218504
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
SMILES OC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 383901
Merck Index 14,6876
Solubility Information Soluble in water (1.7mg/ml),and ammonium hydroxide. Insoluble in chloroform.
Formula Weight 156.10 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Orotic acid hydrate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-619-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.