Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orotidine 5′-monophosphate trisodium | 68244-58-6 | 5 MG
SDP

Catalog No. 5000418578
Encompass_Preferred

Orotidine 5'-monophosphate trisodium is a pyrimidine nucleotide synthesized via the de novo synthesis pathway for DNA synthesis in various microorganisms, including M. tuberculosis, S. cerevisiae, S. typhimurium, and P. falciparum. Its synthesis utilizes phosphoribosyl pyrophosphate (PRPP) and orotic acid (OA) as substrates, catalyzed by orotate phosphoribosyltransferase (OPRT).

  • High purity: 99.0%
  • Synonyms include Orotidine monophosphate trisodium and Orotidylic acid trisodium
  • Molecular weight: 434.14
  • Chemical formula: C10H10N2Na3O11P
  • Appears as a white to off-white solid
  • Soluble in H2O at 50 mg/mL (115.17 mM)
  • Plays a role in DNA synthesis pathway

Catalog No. 50-004-18578 Supplier Medchemexpress LLC Supplier No. HYN8060A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orotidine 5'-monophosphate trisodium is a pyrimidine nucleotide synthesized via the de novo synthesis pathway for DNA synthesis in various microorganisms, including M. tuberculosis, S. cerevisiae, S. typhimurium, and P. falciparum. Its synthesis utilizes phosphoribosyl pyrophosphate (PRPP) and orotic acid (OA) as substrates, catalyzed by orotate phosphoribosyltransferase (OPRT).

  • High purity: 99.0%
  • Synonyms include Orotidine monophosphate trisodium and Orotidylic acid trisodium
  • Molecular weight: 434.14
  • Chemical formula: C10H10N2Na3O11P
  • Appears as a white to off-white solid
  • Soluble in H2O at 50 mg/mL (115.17 mM)
  • Plays a role in DNA synthesis pathway

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.