Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oroxin A | 57396-78-8 | 432.38 | 1 MG
SDP

Catalog No. 5000415904
Encompass_Preferred

Oroxin A is the main component of an ethanol-water extract from *Oroxylum indicum* (L.) Kurz (Bignoniaceae) seed (OISE). It acts as a partial PPARγ agonist, activating PPARγ transcriptional activation by docking into the PPARγ protein ligand-binding domain. Oroxin A also shows inhibitory activity against α-glucosidase and antioxidant capacity. Additionally, Oroxin A exhibits anti-breast cancer effects by inducing ER stress-mediated senescence.

  • Major component of *Oroxylum indicum* seed extract.
  • Partial PPARγ agonist.
  • Activates PPARγ transcriptional activation.
  • Inhibits α-glucosidase.
  • Exhibits antioxidant capacity.
  • Induces ER stress-mediated senescence.
  • Purity: 99.76%.
  • Appearance: Solid, light yellow to yellow.

Catalog No. 50-004-15904 Supplier Medchemexpress LLC Supplier No. HYN20251MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oroxin A is the main component of an ethanol-water extract from *Oroxylum indicum* (L.) Kurz (Bignoniaceae) seed (OISE). It acts as a partial PPARγ agonist, activating PPARγ transcriptional activation by docking into the PPARγ protein ligand-binding domain. Oroxin A also shows inhibitory activity against α-glucosidase and antioxidant capacity. Additionally, Oroxin A exhibits anti-breast cancer effects by inducing ER stress-mediated senescence.

  • Major component of *Oroxylum indicum* seed extract.
  • Partial PPARγ agonist.
  • Activates PPARγ transcriptional activation.
  • Inhibits α-glucosidase.
  • Exhibits antioxidant capacity.
  • Induces ER stress-mediated senescence.
  • Purity: 99.76%.
  • Appearance: Solid, light yellow to yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.