Learn More
Medchemexpress LLC Oroxin A | 57396-78-8 | 432.38 | 1 ML
SDP
Catalog No. 5000416117
Encompass_Preferred
Oroxin A is the major component of an ethanol-water Oroxylum indicum (L.) Kurz (Bignoniaceae) seed extract (OISE). It functions as a partial PPARγ agonist, activating PPARγ transcriptional activity by docking into the PPARγ protein ligand-binding domain.
  • Functions as a partial PPARγ agonist
  • Activates PPARγ transcriptional activity
  • Exhibits inhibitory activity against α-glucosidase
  • Possesses antioxidant capacity
  • Induces anti-breast cancer effects by ER stress-mediated senescence
Catalog No. 50-004-16117 Supplier Medchemexpress LLC Supplier No. HYN202510MM/1ML
Oroxin A is the major component of an ethanol-water Oroxylum indicum (L.) Kurz (Bignoniaceae) seed extract (OISE). It functions as a partial PPARγ agonist, activating PPARγ transcriptional activity by docking into the PPARγ protein ligand-binding domain.
  • Functions as a partial PPARγ agonist
  • Activates PPARγ transcriptional activity
  • Exhibits inhibitory activity against α-glucosidase
  • Possesses antioxidant capacity
  • Induces anti-breast cancer effects by ER stress-mediated senescence