Learn More
Apexbio Technology LLC OROXYLIN-A-1MG
SDP
Catalog No. 5000570731
Encompass_Preferred
Oroxylin A (CAS No 480-11-5) is a naturally occurring flavonoid originally isolated from Scutellaria baicalensis used extensively in biomedical research It functions primarily as an inhibitor of the GABAergic system influencing various neurotransmitter pathways and exhibiting neuroprotective effects In vitro its activity is often observed in the nanomolar to low micromolar range Oroxylin A has been studied for its potential applications in neurodegenerative diseases cancer research and inflammation It is commonly tested in cell models such as cancer cell lines and neuron cultures as well as in animal models like rodents for efficacy and safety profiling This compound is also explored in drug screening processes for its potential therapeutic benefits Experimental concentrations typically depend on the specific research design ensuring optimal efficacy while minimizing adverse effects
Catalog No. 50-005-70731 Supplier Apexbio Technology LLC Supplier No. N28821
Add to Cart
Add to Cart
Oroxylin A (CAS No 480-11-5) is a naturally occurring flavonoid originally isolated from Scutellaria baicalensis used extensively in biomedical research It functions primarily as an inhibitor of the GABAergic system influencing various neurotransmitter pathways and exhibiting neuroprotective effects In vitro its activity is often observed in the nanomolar to low micromolar range Oroxylin A has been studied for its potential applications in neurodegenerative diseases cancer research and inflammation It is commonly tested in cell models such as cancer cell lines and neuron cultures as well as in animal models like rodents for efficacy and safety profiling This compound is also explored in drug screening processes for its potential therapeutic benefits Experimental concentrations typically depend on the specific research design ensuring optimal efficacy while minimizing adverse effects