Learn More
Medchemexpress LLC Oroxylin A | 480-11-5 | 284.26 | 1 MG
SDP
Catalog No. 5000388842
Encompass_Preferred
Oroxylin A is an active flavonoid compound with strong anti-cancer effects. It inhibits the IL-6/STAT3 pathway and NF-κB signaling, inhibits cell proliferation, and induces apoptosis. Oroxylin A also inhibits colitis-related carcinogenesis.
  • Active flavonoid compound
  • Strong anti-cancer effects
  • Inhibits the IL-6/STAT3 pathway and NF-κB signaling
  • Inhibits cell proliferation and induces apoptosis
  • Inhibits colitis-related carcinogenesis
Catalog No. 50-003-88842 Supplier Medchemexpress LLC Supplier No. HYN05601MG
Oroxylin A is an active flavonoid compound with strong anti-cancer effects. It inhibits the IL-6/STAT3 pathway and NF-κB signaling, inhibits cell proliferation, and induces apoptosis. Oroxylin A also inhibits colitis-related carcinogenesis.
  • Active flavonoid compound
  • Strong anti-cancer effects
  • Inhibits the IL-6/STAT3 pathway and NF-κB signaling
  • Inhibits cell proliferation and induces apoptosis
  • Inhibits colitis-related carcinogenesis