Learn More
Medchemexpress LLC Oroxylin A | 480-11-5 | 284.26 | 1 ML
SDP
Catalog No. 5000388769
Encompass_Preferred
Oroxylin A is an active flavonoid compound demonstrating potent anti-cancer effects by inhibiting the IL-6/STAT3 pathway and NF-κB signaling, which suppresses cell proliferation and induces apoptosis. It also inhibits colitis-related carcinogenesis, making it a valuable tool for various research applications.
  • Active flavonoid compound
  • Inhibits IL-6/STAT3 pathway
  • Inhibits NF-κB signaling
  • Suppresses cell proliferation
  • Induces apoptosis
  • Inhibits colitis-related carcinogenesis
  • For research use only
Catalog No. 50-003-88769 Supplier Medchemexpress LLC Supplier No. HYN056010MM/1ML
Oroxylin A is an active flavonoid compound demonstrating potent anti-cancer effects by inhibiting the IL-6/STAT3 pathway and NF-κB signaling, which suppresses cell proliferation and induces apoptosis. It also inhibits colitis-related carcinogenesis, making it a valuable tool for various research applications.
  • Active flavonoid compound
  • Inhibits IL-6/STAT3 pathway
  • Inhibits NF-κB signaling
  • Suppresses cell proliferation
  • Induces apoptosis
  • Inhibits colitis-related carcinogenesis
  • For research use only