Learn More
Medchemexpress LLC Oroxylin A-7-O-glucuronide | 36948-76-2 | 1 ML
SDP
Catalog No. 5000421516
Encompass_Preferred
Oroxylin A-7-O-glucuronide, also known as Oroxyloside, is an orally active flavonoid glucuronide and a metabolite of Oroxylin A. It can be extracted from the dried root of Scutellaria baicalensis. This compound exhibits a range of biological activities:
  • Prolyl oligopeptidase inhibitory activity
  • Inhibits the JNK pathway
  • Upregulates PPARγ
  • Inhibits NF-κB p65 nuclear translocation
  • Reduces cytokine (IL-1β, IL-6) production
  • Exhibits anti-angiogenic, anti-tumor (glioma, liver cancer), anti-inflammatory, and hepatoprotective activities
Catalog No. 50-004-21516 Supplier Medchemexpress LLC Supplier No. HYN248110MM/1ML
Oroxylin A-7-O-glucuronide, also known as Oroxyloside, is an orally active flavonoid glucuronide and a metabolite of Oroxylin A. It can be extracted from the dried root of Scutellaria baicalensis. This compound exhibits a range of biological activities:
  • Prolyl oligopeptidase inhibitory activity
  • Inhibits the JNK pathway
  • Upregulates PPARγ
  • Inhibits NF-κB p65 nuclear translocation
  • Reduces cytokine (IL-1β, IL-6) production
  • Exhibits anti-angiogenic, anti-tumor (glioma, liver cancer), anti-inflammatory, and hepatoprotective activities