Learn More
Medchemexpress LLC Oroxylin A-7-O-glucuronide | 36948-76-2 | MFCD23379930 | 99.8% | 460.39 g/mol | C22H20O11 | 10 MG
SDP
Catalog No. 5000289858
Encompass_Preferred
Oroxylin A-7-O-glucuronide is an orally active flavonoid glucuronide and a metabolite of oroxylin A supplied as an analytical research standard. It is used in biochemical and pharmacological studies and has reported activity as a prolyl oligopeptidase inhibitor and a modulator of JNK, NF-κB, and PPARγ signaling, with observed anti-inflammatory, anti-angiogenic, anti-tumor, and hepatoprotective effects.
  • High purity (99.8%) analytical standard suitable for in vitro and in vivo research
  • Chemical identity: C22H20O11; molecular weight 460.39 g/mol
  • Supplied in small-mass vials appropriate for assay development and analytical workflows
  • Soluble in DMSO for easy stock solution preparation
  • Accompanied by datasheet and safety data sheet for handling and regulatory information
Catalog No. 50-002-89858 Supplier Medchemexpress LLC Supplier No. HYN248110MG
Oroxylin A-7-O-glucuronide is an orally active flavonoid glucuronide and a metabolite of oroxylin A supplied as an analytical research standard. It is used in biochemical and pharmacological studies and has reported activity as a prolyl oligopeptidase inhibitor and a modulator of JNK, NF-κB, and PPARγ signaling, with observed anti-inflammatory, anti-angiogenic, anti-tumor, and hepatoprotective effects.
  • High purity (99.8%) analytical standard suitable for in vitro and in vivo research
  • Chemical identity: C22H20O11; molecular weight 460.39 g/mol
  • Supplied in small-mass vials appropriate for assay development and analytical workflows
  • Soluble in DMSO for easy stock solution preparation
  • Accompanied by datasheet and safety data sheet for handling and regulatory information