Learn More
TARGETMOL CHEMICALS INC Oroxylin A-7-O-glucuronide 5MG
SDP
Catalog No. 502880460
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Oroxylin A-7-O-glucuronide (Oroxylin A Glucoronide) and wogonoside should also be served as the chemical markers together with baicalin for the quality control of herbs and proprietary traditional Chinese medicine (PTCM) products of radix Scutellariae. Oroxylin A 7-O-glucuronide, Oroxin A, and Oroxin B can bind to the "stem" region of alpha-hemolysin (alpha-HL), when inhibiting the hemolytic activity of alpha-HL. Oroxylin A 7-O-glucuronide and 4', 5, 7-trihydroxy-6-methoxyflavone at a concentration of 100 microM inhibit 34 and 43% of total prolyl oligopeptidase (POP) activity, respectively. Purity 99.3%
Catalog No. 50-288-0460 Supplier TARGETMOL CHEMICALS INC Supplier No. T38915MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Oroxylin A-7-O-glucuronide (Oroxylin A Glucoronide) and wogonoside should also be served as the chemical markers together with baicalin for the quality control of herbs and proprietary traditional Chinese medicine (PTCM) products of radix Scutellariae. Oroxylin A 7-O-glucuronide, Oroxin A, and Oroxin B can bind to the "stem" region of alpha-hemolysin (alpha-HL), when inhibiting the hemolytic activity of alpha-HL. Oroxylin A 7-O-glucuronide and 4', 5, 7-trihydroxy-6-methoxyflavone at a concentration of 100 microM inhibit 34 and 43% of total prolyl oligopeptidase (POP) activity, respectively. Purity 99.3%