Learn More
YSI 3P LifeScience ORP BUFFR SOLN.PH 7,RH 28
Catalog No. NC0616112
Encompass_Preferred
ORP buffer solution pH 7, UH = 427 mV (CH), 1 bottle with 250 ml.
Catalog No. NC0616112 Supplier YSI 3P LifeScience Supplier No. 109740
ORP buffer solution pH 7, UH = 427 mV (CH), 1 bottle with 250 ml.