Learn More
Grainger ORP Electrode CPVC BNR Flat Tip
Catalog No. 502767634
Encompass_Preferred
The SENSOREX S265CD-ORP ORP Electrode is an oxidation-reduction potential (ORP) electrode designed for water treatment applications. It has a platinum sensor and ceramic junction for measuring ORP in the range of ±2000 mV. The electrode has a 3/4" NPT thread connection, epoxy body, and 10 ft cable. It requires a separate reference electrode and is compatible with SENSOREX ORP controllers and transmitters. The electrode provides stable ORP measurement in treated water.
Catalog No. 50-276-7634 Supplier Grainger Supplier No. 16Z020
The SENSOREX S265CD-ORP ORP Electrode is an oxidation-reduction potential (ORP) electrode designed for water treatment applications. It has a platinum sensor and ceramic junction for measuring ORP in the range of ±2000 mV. The electrode has a 3/4" NPT thread connection, epoxy body, and 10 ft cable. It requires a separate reference electrode and is compatible with SENSOREX ORP controllers and transmitters. The electrode provides stable ORP measurement in treated water.