Learn More
Grainger ORP Electrode Epoxy BNC Extended Tip
Catalog No. 502768162
Encompass_Preferred
The Grainger Approved ORP Electrode Epoxy BNC is an oxidation reduction potential electrode with a BNC connector. It has a 3/4 inch diameter epoxy body and platinum sensor. The electrode measures ORP in water treatment, pools, spas, industrial processes and environmental monitoring. It has an operating temperature range of 0 to 80°C. The cable is 6 feet long.
Catalog No. 50-276-8162 Supplier Grainger Supplier No. 16Y972
The Grainger Approved ORP Electrode Epoxy BNC is an oxidation reduction potential electrode with a BNC connector. It has a 3/4 inch diameter epoxy body and platinum sensor. The electrode measures ORP in water treatment, pools, spas, industrial processes and environmental monitoring. It has an operating temperature range of 0 to 80°C. The cable is 6 feet long.