Learn More
Grainger ORP Electrode Epoxy BNC Extended Tip
Catalog No. 502768174
Encompass_Preferred
The Grainger Approved ORP Electrode is an oxidation reduction potential electrode used to measure ORP in water. It has a polypropylene body, platinum sensor, BNC connector, and 3/4" NPT threads. The electrode has a measuring range of ±2000 mV with ±15 mV accuracy. It is compatible with controllers and monitors that accept BNC connections and is rated for use in temperatures up to 80°C. The electrode requires calibration and storage solution for proper maintenance. Overall, this is an industrial-grade ORP electrode for water quality monitoring and control applications.
Catalog No. 50-276-8174 Supplier Grainger Supplier No. 16Y973
The Grainger Approved ORP Electrode is an oxidation reduction potential electrode used to measure ORP in water. It has a polypropylene body, platinum sensor, BNC connector, and 3/4" NPT threads. The electrode has a measuring range of ±2000 mV with ±15 mV accuracy. It is compatible with controllers and monitors that accept BNC connections and is rated for use in temperatures up to 80°C. The electrode requires calibration and storage solution for proper maintenance. Overall, this is an industrial-grade ORP electrode for water quality monitoring and control applications.