Learn More
Grainger ORP Electrode Epoxy BNC Extended Tip
Catalog No. 502765156
Encompass_Preferred
The SensoRex ORP Electrode measures ORP in water treatment. It features a polypropylene body, platinum element, and BNC connector. With a range of ±2000 mV and accuracy of ±5 mV, it operates from 0-80°C. A separate reference electrode and meter are needed. It responds in <10 seconds and works with oxidizing/reducing chemicals. Suitable for pools, spas, industrial, and drinking water applications, it requires regular calibration and cleaning. It comes with a 12-month warranty.
Catalog No. 50-276-5156 Supplier Grainger Supplier No. 16Y974
The SensoRex ORP Electrode measures ORP in water treatment. It features a polypropylene body, platinum element, and BNC connector. With a range of ±2000 mV and accuracy of ±5 mV, it operates from 0-80°C. A separate reference electrode and meter are needed. It responds in <10 seconds and works with oxidizing/reducing chemicals. Suitable for pools, spas, industrial, and drinking water applications, it requires regular calibration and cleaning. It comes with a 12-month warranty.