Learn More
Grainger ORP Electrode Epoxy BNC Polished Tip
Catalog No. 502767672
Encompass_Preferred
The Grainger 16Y971 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning, scouring, and finishing. It features an open web construction with non-scratching fibers that are ideal for cleaning pots, pans, grills, tiles, fiberglass, and more. This 6 inch by 9 inch hand pad has a medium duty abrasiveness and is latex-free and biodegradable. It effectively removes grease, grime, rust stains, and more from a variety of surfaces. The Scotch-Brite General Purpose Hand Pad from 3M is a versatile cleaning tool.
Catalog No. 50-276-7672 Supplier Grainger Supplier No. 16Y971
The Grainger 16Y971 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning, scouring, and finishing. It features an open web construction with non-scratching fibers that are ideal for cleaning pots, pans, grills, tiles, fiberglass, and more. This 6 inch by 9 inch hand pad has a medium duty abrasiveness and is latex-free and biodegradable. It effectively removes grease, grime, rust stains, and more from a variety of surfaces. The Scotch-Brite General Purpose Hand Pad from 3M is a versatile cleaning tool.