Learn More
Grainger ORP Electrode Epoxy BNR Round Tip
Catalog No. 502767986
Encompass_Preferred
The SENSOREX S222CD-ORP ORP Electrode is an oxidation reduction potential (ORP) electrode designed for water treatment applications. It has a platinum sensor and ceramic reference junction with a double junction design for extended service life. The electrode has a 3/4" NPT thread connection and measures 12" length x 1" diameter. It has a temperature range of 0-80°C, pressure range of 0-100 psig, and pH range of 0-14. The S222CD-ORP provides accurate ORP measurement for monitoring water treatment processes.
Catalog No. 50-276-7986 Supplier Grainger Supplier No. 16Y963
The SENSOREX S222CD-ORP ORP Electrode is an oxidation reduction potential (ORP) electrode designed for water treatment applications. It has a platinum sensor and ceramic reference junction with a double junction design for extended service life. The electrode has a 3/4" NPT thread connection and measures 12" length x 1" diameter. It has a temperature range of 0-80°C, pressure range of 0-100 psig, and pH range of 0-14. The S222CD-ORP provides accurate ORP measurement for monitoring water treatment processes.