Learn More
Cole-Parmer Oakton ORP electrode; single-junction, gel-filled, BNC connector SKU 5900175
Catalog No. NC9414641 Shop All Cole-Parmer Products
Encompass_Preferred
Combining durability with reliability, the electrode's epoxy-body housing resists impact. Ideal for general purpose lab and field applications. Compatible with most brands of pH meters.
Catalog No. NC9414641 Supplier Cole-Parmer Supplier No. 5900175
Combining durability with reliability, the electrode's epoxy-body housing resists impact. Ideal for general purpose lab and field applications. Compatible with most brands of pH meters.