Learn More
Hach Company ORP Encap Sens-LCP-Convert-5
Catalog No. 5000207614 Shop All Hach Company Products
Encompass
ORP Encapsulated Sensor-LCP Body-Convertible-5 Wire-Platinum Electrode Material-30-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 9m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode
Catalog No. 50-002-07614 Supplier Hach Company Supplier No. 2028R030
ORP Encapsulated Sensor-LCP Body-Convertible-5 Wire-Platinum Electrode Material-30-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 9m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode