Learn More
Hach Company ORP Encap Sens-LCP-Union-2Wr
Catalog No. 5000207996 Shop All Hach Company Products
Encompass
ORP Encapsulated Sensor-LCP Body-2 Wire-Union Mount-10-ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter Platinum Electrode
Catalog No. 50-002-07996 Supplier Hach Company Supplier No. 2458R0
ORP Encapsulated Sensor-LCP Body-2 Wire-Union Mount-10-ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter Platinum Electrode