Learn More
Hach Company Intellical™ MTC101 Laboratory Low Maintenance Gel Filled ORP/RedOx Electrode, 3 m Cable
Catalog No. 501998054 Shop All Hach Company Products
Encompass
Intellical™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) electrode with a non-refillable, gel-electrolyte reference and built-in temperature sensor. MTC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. The laboratory version of this metallic electrode is shockproff with its epoxy plastic body protecting down to the platinum sensing element. The Electrode is available with a 3 or 1 m cable. The MTC101 electrode is ideal for measuring ORP/RedOx absolute mV values in wastewater, drinking water and aqueous media applications. The MTC101 is not suitable for use with organic solvents.
Catalog No. 50-199-8054 Supplier Hach Company Supplier No. MTC10103
Intellical™ MTC101 is a digital, combination oxidation reduction potential (ORP/Redox) electrode with a non-refillable, gel-electrolyte reference and built-in temperature sensor. MTC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. The laboratory version of this metallic electrode is shockproff with its epoxy plastic body protecting down to the platinum sensing element. The Electrode is available with a 3 or 1 m cable. The MTC101 electrode is ideal for measuring ORP/RedOx absolute mV values in wastewater, drinking water and aqueous media applications. The MTC101 is not suitable for use with organic solvents.