Learn More
Grainger ORP Insertion Assembly CPVC BNC Flat Tip
Catalog No. 502767497
Encompass_Preferred
The SENSOREX S268CD-ORP ORP Electrode is an oxidation-reduction potential (ORP) electrode designed for water treatment applications. It has a platinum measuring element and a double junction reference with a ceramic pin junction and sleeve junction. The electrode has a polypropylene body, epoxy head, and BNC connector. It has a measurement range of ±2000 mV with a 0 to 80°C temperature range. The S268CD-ORP provides accurate ORP measurements in treated water to monitor disinfection and oxidation processes.
Catalog No. 50-276-7497 Supplier Grainger Supplier No. 16Z022
The SENSOREX S268CD-ORP ORP Electrode is an oxidation-reduction potential (ORP) electrode designed for water treatment applications. It has a platinum measuring element and a double junction reference with a ceramic pin junction and sleeve junction. The electrode has a polypropylene body, epoxy head, and BNC connector. It has a measurement range of ±2000 mV with a 0 to 80°C temperature range. The S268CD-ORP provides accurate ORP measurements in treated water to monitor disinfection and oxidation processes.