Learn More
Hach Company Intellical™ MTC301 Laboratory General Purpose Refillable ORP/RedOx Electrode, 3 m Cable
Catalog No. 501998060 Shop All Hach Company Products
Encompass
Intellical™ MTC301 is a digital combination oxidation reduction potential (ORP/RedOx) electrode with built-in temperature sensor. The MTC301 has a refillable with a ceramic pin reference junction. The laboratory version of this metallic electrode is shockproff with its epoxy plastic body protecting down to the platinum sensing element. The Electrode is available with a 3 or 1 m cable. The MTC301 electrode is ideal for measuring ORP/RedOx absolute mV values in wastewater, drinking water and aqueous media applications. The MTC101 is not suitable for use with organic solvents. A 59 mL bottle of reference electrolyte filling solution (3M KCl solution saturated with AgCl) is included with the probe.
Catalog No. 50-199-8060 Supplier Hach Company Supplier No. MTC30103
Intellical™ MTC301 is a digital combination oxidation reduction potential (ORP/RedOx) electrode with built-in temperature sensor. The MTC301 has a refillable with a ceramic pin reference junction. The laboratory version of this metallic electrode is shockproff with its epoxy plastic body protecting down to the platinum sensing element. The Electrode is available with a 3 or 1 m cable. The MTC301 electrode is ideal for measuring ORP/RedOx absolute mV values in wastewater, drinking water and aqueous media applications. The MTC101 is not suitable for use with organic solvents. A 59 mL bottle of reference electrolyte filling solution (3M KCl solution saturated with AgCl) is included with the probe.