Learn More
Hach Company ORP Sens-LCP-Convert-2 Wire
Catalog No. 5000207541 Shop All Hach Company Products
Encompass
ORP Sensor-LCP Body-Convertible-2 Wire-Platinum Electrode-10-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter Platinum Electrode
Catalog No. 50-002-07541 Supplier Hach Company Supplier No. 2428R0
ORP Sensor-LCP Body-Convertible-2 Wire-Platinum Electrode-10-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter Platinum Electrode