Learn More
Hach Company ORP Sens-LCP-Convert-5 Wire
Catalog No. 5000207615 Shop All Hach Company Products
Encompass
ORP Sensor-LCP Body-Convertible-5 Wire-50-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 15m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode
Catalog No. 50-002-07615 Supplier Hach Company Supplier No. 2028R050
ORP Sensor-LCP Body-Convertible-5 Wire-50-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 15m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode