Learn More
Hach Company ORP Sens-LCP-Convert-5 Wire
Catalog No. 5000207657 Shop All Hach Company Products
Encompass
ORP Sensor-LCP Body-Convertible-5 Wire-25-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 7 5m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode
Catalog No. 50-002-07657 Supplier Hach Company Supplier No. 2028R025
ORP Sensor-LCP Body-Convertible-5 Wire-25-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 7 5m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode