Learn More
Hach Company ORP Sens-LCP-Convert-5 Wire
Catalog No. 5000208004 Shop All Hach Company Products
Encompass
ORP Sensor-LCP Body-Convertible-5 Wire-Platinum ElectrodeLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode
Catalog No. 50-002-08004 Supplier Hach Company Supplier No. 2028R0
ORP Sensor-LCP Body-Convertible-5 Wire-Platinum ElectrodeLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode