Learn More
Hach Company ORP Sens-LCP-Convert-Gold-5
Catalog No. 5000207702 Shop All Hach Company Products
Encompass
ORP Sensor-LCP Body-Convertible-Gold Electrode Material-5 Wire-10-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Gold Electrode
Catalog No. 50-002-07702 Supplier Hach Company Supplier No. 2028R1
ORP Sensor-LCP Body-Convertible-Gold Electrode Material-5 Wire-10-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Gold Electrode