Learn More
Hach Company ORP Sens-PPS-Conver-5 Wire
Catalog No. 5000208003 Shop All Hach Company Products
Encompass
ORP Sensor-PPS Body-Convertible-5 Wire-Platinum Electrode Material-10-ft CablePPS Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode
Catalog No. 50-002-08003 Supplier Hach Company Supplier No. 2022R0
ORP Sensor-PPS Body-Convertible-5 Wire-Platinum Electrode Material-10-ft CablePPS Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Platinum Electrode